missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ OSGEP Overexpression Lysate

Product Code. 18298643 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
Product Code. Quantity unitSize
18298643 0.1 mg 0.1mg
1 options
This item is not returnable. View return policy

Product Code. 18298643

Brand: Novus Biologicals™ NBL113999

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
The OSGEP Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Cell Line The lysate was created in HEK293T cells, using plasmid ID RC2789 and based on accession number NM_1787. The protein contains a C-terminal DDK tag.
For Use With (Application) Western Blot
Quantity 0.1 mg
Storage Buffer RIPA buffer
Purity Protein
Test Specificity Homo sapiens O-sialoglycoprotein endopeptidase (OSGEP), mRNA.
Content And Storage -80°C. Avoid freeze-thaw cycles.
Protein State Native

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.