Learn More
BINDER™ Overheating Alarm Function for Drying and Heating Chambers
Product Code. 16434169 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
BINDER Chambers ED 23
BINDER Chambers FD 23
BINDER Chambers FED 400
Unit Size:
Each
Product Code. 16434169
Brand: BINDER™ 80121299
Only null left
Add to basket
Add to basket
The Overheating Alarm Function for Drying and Heating Chambers features a switchable acoustic alarm, with adjustable limit value on the independent temperature safety device.
  • Audible alarm function for overheating
  • Features switchable acoustic alarm
  • Features adjustable limit value
  • Designed for use with BINDER™ Drying and Heating Chambers
Specifications
Description Alarm function for overheating - Switchable acoustic alarm, with adjustable limit value on the independent temperature safety device
For Use With (Equipment) BINDER Chambers ED 23
Type Adjustable Overheating Alarm
Includes Switchable acoustic alarm
Voltage 230 V