missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ CAT Supplement
The Oxoid CAT Supplement isolates Campylobacter spp.
195.00€ - 675.00€
Specifications
Species | Campylobacter |
---|---|
Form | Freeze-dried |
Product Type | Microbiology Media |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12742020
|
Thermo Scientific™
SR0174H |
For 2L medium |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
11434219
|
Thermo Scientific™
SR0174E |
10/Pk. |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Description
Oxoid CAT Supplement is used for the isolation of Campylobacter spp., especially C. upsaliensis or Acrobacter.
- Add to Campylobacter Blood-Free Selective Agar Base (CCDA Preston), Part No. CM0739B for the isolation of Campylobacter spp.
- Add to Arcobacter Broth Base, Part No. CM0965B for the isolation of Arcobacter spp.
Specifications
Campylobacter | |
Freeze-dried | |
Microbiology Media |