Learn More
Thermo Scientific™ EC Broth (Dehydrated)
Description
Selectively detect Escherichia coli from food and environmental samples with this medium.
Selectively detect Escherichia coli from food and environmental samples with Thermo Scientific™ Oxoid™ EC Broth (Dehydrated). EC Broth is a selective medium for the differentiation of fecal coliforms and the confirmatory test for Escherichia coli from food and environmental samples1,2. It is also suitable for the enumeration of presumptive E. coli in milk and milk products using the most probable number technique3.
Escherichia coli is a common Gram-negative micro-organism which may be present in food and water samples. The presence of E.coli cells may be indicative of fecal contamination, although their ingestion does not necessarily have adverse effects on health. Nevertheless, certain strains are responsible for diarrhoeal disease and can also lead to more serious forms of illness.
EC Broth has shown to be a reliable medium producing dependable results in 24 to 48 hours3.
- Selective: the addition of bile salts no.3 in the medium is inhibitory to gram-positive bacteria especially bacilli and fecal Streptococci making the medium selective for Escherichia coli
- Easy-to-read: gas production as a result of lactose fermentation is a presumptive evidence of the presence of coliform bacteria.
The presence of E. coli in water is also regarded as a specific indicator of fecal pollution, but lack of rapid identification methods means that the non-specific group - fecal coliforms - is also used as the indicator of such pollution. In this regard, it is important to differentiate between coliforms of fecal origin and those from other sources.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
Specifications
| Certifications/Compliance | Industrial Reference: ISO, NMKL standards. |
| Description | EC Broth, Modified |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| Quantity | 500 g |
| Yield | For 13.5L medium |
For Laboratory Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.