Learn More
Thermo Scientific™ Oxoid™ Lab-Lemco Beef Extract Powder
Description
Thermo Scientific™ Oxoid Lab-Lemco Powder is a Beef Extract made from specially selected raw materials for quality and bio-logical safety.
Use with other refined ingredients, e.g. Neutralized bacteriological peptone (LP0034) to prepare culture media which do not require filtration.
May be used in fermentation processes.
Industrial Reference: NMKL
- May be used in fermentation processes
- Industrial Reference: NMKL
Specifications
Specifications
| Content And Storage | Dehydrated Powder, Ambient & Dry Storage |
| Form | Dehydrated Powder |
| Product Type | Peptone |
| For Use With (Application) | Fermentation processes |
| Quantity | 500 g |
| Protein Source | Beef |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.