Learn More
Thermo Scientific™ Oxoid™ Neomycin Selective Supplement
Product Code. 13225399
Change view
Click to view available options
Quantity:
10 x 150 mg
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13225399 10 x 150 mg Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13225399 Supplier Thermo Scientific™ Supplier No. SR0163H
The Oxoid Neomycin Selective Supplement (150mg/vial) allows the growth of clostridia and other anaerobes.
Oxoid Neomycin Selective Supplement (150mg/vial) allows the growth of clostridia and other anaerobes.
  • Add to Blood Agar, Wilkins-Chalgren Anaerobe Agar, Part No. CM0619B, or Columbia Blood Agar, Part No. CM0331B
  • Each vial supplements 2L of medium
TRUSTED_SUSTAINABILITY
Specifications
Description Neomycin Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 10 x 150 mg
Yield Each vial for 2L medium