Learn More
Thermo Scientific™ Tryptone Glucose Extract Agar (Dehydrated)
Product Code. 10700544
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10700544 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10700544 Supplier Thermo Scientific™ Supplier No. CM0127B
Enumerate bacteria in water and dairy products and detect thermophilic organisms with this medium.
Enumerate bacteria in water and dairy products and detect thermophilic organisms with Thermo Scientific™ Oxoid™ Tryptone Glucose Extract Agar (Dehydrated). The medium is recommended for the plate count of water and dairy products1,2. It can also be used for the detection of thermophilic bacteria in dairy products.
Use Tryptone Glucose Extract Agar for enumeration of water and dairy products and for the detection of thermophilic organisms.
The Standard Methods Committee of the American Public Health Association recommended that sterile milk should be added to this medium only when the dilution of the original specimen was greater than 1 in 10. The addition of 10mL of a sterile 10% solution of Skim Milk Powder LP0031 per liter is suitable for this purpose.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial Reference: AOAC.
Description Tryptone Glucose Extract Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 20.8L medium
For Laboratory Use Only