Learn More
Welch™ P 8 E Rotary Vacuum Pump
Brand:  Welch™ 321004
Additional Details : Weight : 25.00000kg
Product Code. 10084514
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Specifications
120/143 L/min.
150 mm
<49 dB
21 kg
215 mm
Rotary Vane Vacuum Pump
100/115/230 V 50/60 Hz
P 8 E
200 w
50/60 Hz
405 mm
100/115/230 V