Learn More
p-Terphenyl, 99+%
Product Code. 11434010
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
Product Code. 11434010
Brand: Thermo Scientific Alfa Aesar A14833.30
Request Bulk or Custom Format
CAS: 92-94-4 | C18H14 | 230.31 g/mol
p-Terphenyl is a specialty material, and may be used in ionized radiation detectors, nonpolar laser dye, and single molecule optical probe of scanning near-field microscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
p-Terphenyl is a specialty material, and may be used in ionized radiation detectors, nonpolar laser dye, and single molecule optical probe of scanning near-field microscopy.
Solubility
Insoluble in water. Soluble in hot benzene. Very soluble in hot ethyl alcohol. 
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 92-94-4
Molecular Formula C18H14
Molecular Weight (g/mol) 230.31
MDL Number MFCD00003061
InChI Key XJKSTNDFUHDPQJ-UHFFFAOYSA-N
Synonym p-terphenyl, 1,1':4',1-terphenyl, p-diphenylbenzene, p-triphenyl, santowax p, 4-phenylbiphenyl, 4-phenyldiphenyl, para-terphenyl, biphenyl, 4-phenyl, pyrogallol tannin
PubChem CID 7115
ChEBI CHEBI:52242
IUPAC Name 1,4-diphenylbenzene
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=CC=C3
Specifications
Melting Point 212°C to 215°C
Density 1.23
Boiling Point 405°C
Flash Point 207°C (404°F)
Quantity 250 g
Beilstein 1908447
Solubility Information Insoluble in water. Soluble in hot benzene. Very soluble in hot ethyl alcohol.
Formula Weight 230.31
Percent Purity ≥99%
Chemical Name or Material p-Terphenyl