Learn More
Sartorius Polyethersulfone Membrane Filters
Shop All Sartorius Products
Click to view available options
Diameter (Metric):
25 mm
47 mm
50 mm
Pore Size:
0.1 μm
0.2 μm
0.5 μm
Unit Size:
Pack of 100
Description
Polyethersulfone (PES) filters have a high internal porosity.
They perform well at high flux with an excellent throughput of aqueous solutions over the entire pH range of 1-14. Thanks to their low non-specific protein adsorption the PES type is recommended for filtering biological and pharmaceutical solutions.
Specifications
Specifications
| Disposable | Single-use |
| Filter Type | Membrane Filter |
| Format | Disc |
| Material | PES |
| Material (Membrane) | Polyethersulfone |
| Pore Size | 0.5 μm |
| Sterility | Non-sterile |
| For Use With (Application) | Recommended for filtering biological and pharmaceutical solutions. |
| Quantity | 100/Pk. |
| Thickness (Metric) | 150 μm |
| Show More |