Learn More
Dostmann Electronic P750 Temperature-Humidity Thermometer
Robust, reliable, precise multi-function instruments that are outstanding for their high accuracy of ±0.03°C.
Brand:  Dostmann Electronic 5000-0750
Additional Details : Weight : 0.30000kg
Product Code. 10219321
  • 680.00€ / Each
Estimated Shipment: 03-05-2024
to see stock.
Récupel-bijdrage uitgesloten
Description
Description
  • P750 Temperature-humidity instrument
  • High accurate 1-channel hand ­held ther­mo­meter, max. accuracy + - 0,03°C (PT100)
  • USB interface
  • Optional DE-Graph Windows Software for graphic and tabular documentation
  • High measuring accuracy
  • Integrated calibration function for simple compensation of sensor tolerances