Learn More
Bryson Packaging™ Aluminum Foil
Product Code. 12309558
Click to view available options
Width (Metric):
100mm
500mm
Unit Size:
Each
Product Code. 12309558
Brand: Bryson Packaging™ AKL310040C
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Suited for general wrapping purposes and many laboratory applications. Bryson Packaging™ Aluminum Foil is ideal for use in autoclaves. Available in 100mm wide, 18μM guage and 500mm wide, 12μM guage rolls. Each roll has 75m of quality packaging foil.
  • Suitable for general wrapping purposes and many laboratory applications, including:
    • sealing bottles
    • culture tubes
    • stopping leaks
    • reducing evaporation or heat losses
    • covering corks
    • making weighing boats or dishes
  • 75m rolls available in a variety of widths and gaugess
  • Ideal for use in autoclaves
  • Supplied in a cutter box
Specifications
Applications Autoclavable
Length (English) 246.1 ft.
Color Metallic Silver
Product Type Aluminum Foil in Cutter Box
Width (Metric) 100mm
Material Aluminum
Thickness 18 μm