missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PALCAM Agar Base (Dehydrated)

Product Code. p-200112448
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10056352

Brand: Thermo Scientific™ CM0877B

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Detect and isolate Listeria monocytogenes from food samples with this selective medium.

Detect and isolate Listeria monocytogenes from food samples with Thermo Scientific™ Oxoid™ PALCAM Agar Base (Dehydrated). The PALCAM Agar Base is highly selective and contains double indicator system that allows easy differentiation of Listeria monocytogenes, based upon aesculin fermentation, lack of mannitol fermentation and resultant pH change.

Listeria monocytogenes is the most common pathogenic Listeria spp. and has been shown to be pathogenic to both humans and animals. PALCAM Medium is based on the formulation described by Van Netten et al.1 and is recommended for the isolation of Listeria monocytogenes from foods.

  • Selective: the addition of lithium chloride, ceftazidime, polymyxin B and acriflavine hydrochloride make the medium highly selective for Listeria monocytogenes.
  • Easy-to-read: monocytogenes forms brown/black colonies with a black halo while mannitol positive accompanying flora, such as staphylococci and enterococci, grow as yellow colonies.

PALCAM Medium allows easy differential diagnosis of L. monocytogenes by utilizing the double indicator system.

  1. Aesculin and ferrous iron
  2. Mannitol and phenol red

Listeria monocytogenes hydrolyses aesculin resulting in the formation of a black halo around colonies. L. monocytogenes does not ferment mannitol so can be easily differentiated from contaminants, such as enterococci and staphylococci, which ferment mannitol producing a change from red to yellow in the pH indicator phenol red.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AFNOR, IDF, NMKL.
Description PALCAM Agar Base
Form Powder
Product Type Agar
Quantity 500 g
Yield For 7.2L medium
compliance-icons
Product Identifier
  • PALCAM AGAR BASE
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.
Supplemental information
  • MIXTURE LIST-Contains : Lithium chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.