Learn More
Alfa Aesar™ Palladium, 2.5% Platinum, 2.5% on carbon paste, Type 122
Brand: Alfa Aesar™ H36548.14
Additional Details : CAS : 7440-05-3 Weight : 0.02500kg
Quantity | 25g |
---|
Chemical Identifiers
7440-05-3 | |
106.42 | |
KDLHZDBZIXYQEI-UHFFFAOYSA-N | |
23938 | |
palladium |
Pd | |
MFCD03613605 | |
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon | |
CHEBI:33363 | |
[Pd] |
Specifications
Palladium, 2.5% Platinum | |
7440-05-3 | |
MFCD03613605 | |
KDLHZDBZIXYQEI-UHFFFAOYSA-N | |
palladium | |
23938 | |
Solid | |
Black |
2.5% on carbon paste, Type 122 | |
Pd | |
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon | |
[Pd] | |
106.42 | |
CHEBI:33363 | |
Odorless | |
25g |