Learn More
Alfa Aesar™ Palladium, 5% on carbon paste, Type 761
Brand: Alfa Aesar™ H37526.22
Additional Details : CAS : 7440-05-3 Weight : 0.10000kg
Quantity | 100g |
---|
Chemical Identifiers
7440-05-3 | |
106.42 | |
KDLHZDBZIXYQEI-UHFFFAOYSA-N | |
23938 | |
palladium |
Pd | |
MFCD03457879 | |
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon | |
CHEBI:33363 | |
[Pd] |
Specifications
Palladium | |
7440-05-3 | |
MFCD03457879 | |
KDLHZDBZIXYQEI-UHFFFAOYSA-N | |
palladium | |
23938 | |
Solid | |
Gray |
5% on carbon paste, Type 761 | |
Pd | |
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon | |
[Pd] | |
106.42 | |
CHEBI:33363 | |
Odorless | |
100g |