missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium Standard: Pd @ 1000 μg/mL in 20% HCl, VHG™
Description
Palladium Standard: Pd @ 1000 μg/mL in 20% HCl
Specifications
Specifications
| Chemical Name or Material | Palladium |
| CAS | 5-3-7440 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Palladium |
| Shipping Condition | Room Temperature |
| Molecular Formula | Pd |
| Certifications/Compliance | ISO 17034 |
| Synonym | Palladium on carbon, Palladium Black, Paladio, Palladium/carbon, Palladium, IG 0218A, MPP 030, MPP 050, MPP 080, P 50, P 50 (metal), Palladex 600, Palladium (nanoparticles), Palladium (powder), Palladium element, SFP 1001P, TCP 211 |
| InChI Formula | InChI=1S/Pd |
| SMILES | [Pd] |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?