Learn More
Cole-Parmer™ Parker Hannifin™ P-P-6 Inserts for Soft Tubing
Click to view available options
Tubing Size:
1/4 in.
Unit Size:
Pack of 10
Description
- Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal
- Compression fittings are an excellent choice for connecting rigid tubing systems
- Very good chemical resistance
Specifications
Specifications
| Description | Compression Connection |
| Autoclavable | Not Autoclavable |
| Material | Polypropylene |
| Tubing Size | 1/4 in. |
| Hose Barb | 1/4 in. |
| Outlet Port | 3/8 in. |
| Port Size | 1/4 in. |
| Sterilization Method | Ethylene Oxide |
| Color | White |
| Product Type | Inserts for Soft Tubing |