Learn More
Cole-Parmer™ Parker Hannifin™ PFA Compression Bulkhead Union
Product Code. 11750399
Click to view available options
Tubing Size:
1/4 in.
1/8 in.
Unit Size:
Each
Product Code. 11750399
Brand: Cole-Parmer™ 3132083
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings
  • Superior chemical resistance with durable PFA composition.
  • Get a tight seal at higher pressures.
  • Withstand higher pressures than barbed fittings.
  • Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Specifications
Description Compression Connection
Autoclavable Autoclavable
Material PFA
Tubing Size 1/4 in.
Outlet Port 1/4 x 1/4 in.
Sterilization Method Autoclave or Ethylene Oxide
Color Clear
Product Type Compression Bulkhead Union