Learn More
Tocris Bioscience™ Paroxetine maleate
GREENER_CHOICE
Product Code. 18083782
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 18083782
Brand: Tocris Bioscience™ 2141/10
Highly potent and selective 5-HT uptake inhibitor
Paroxetine maleate is a highly potent and selective 5-HT uptake inhibitor that binds with high affinity to the serotonin transporter (Ki = 0.05 nM). Ki values are 1.1, 350 and 1100 nM for inhibition of [3H]-5-HT, [3H]-l-NA and [3H]-DA uptake respectively. Displays minimal affinity for α1-, α2- or β-adrenoceptors, 5-HT2A, 5-HT1A, D2 or H1 receptors at concentrations below 1000 nM, however displays weak affinity for muscarinic ACh receptors (Ki = 42 nM). Antidepressant and anxiolytic in vivo.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10 mg
Product Type Paroxetine maleate
Purity >99%
Molecular Weight (g/mol) 445.44
Inhibitors Paroxetine maleate