Learn More
Alla France™ BIOTEMP™ Thermometer
Use these mercury free thermometers for a variety of applications. Alla France™ BIOTEMP™ Thermometers feature a yellow back glass with a safe, blue organic filling liquid.
Brand: Alla France™ 5122P150-qp
5.84 EUR valid until 2024-05-31
Use promo code "21638" to get your promotional price.
Additional Details : Weight : 0.02700kg
Description
- Diameter: 6 to 7mm
- Easy to read blue graduation printed and fused in the glass
- Available in partial or total immersion
- A strong triangular ring is molded at the top of the thermometer avoiding the thermometer to roll on an inclined lab table
- Individually packed in recyclable sturdy plastic concertina box's, ideal for storing the instrument when not in use
- Some references are also available in sealed blister packs of six
- Systematically delivered with a Precision and Traceability Certificate
Specifications
305 mm |