Learn More
Thermo Scientific™ Particle-Certified Glass Containers
Product Code. 16875781
Click to view available options
Capacity (Metric):
60 mL
125 mL
250 mL
500 mL
1000 mL
2500 mL
4000 mL
Product Type:
Boston Round Bottle
Jug
Packer Bottle
Sample Jar
Color:
Amber
Clear
No. per Case:
4
12
24
Unit Size:
Each
Pack of 12
Pack of 24
Pack of 4
Product Code. 16875781
Brand: Thermo Scientific™ 13216C/LP
Only null left
Add to basket
Add to basket
Protect the quality of your products or laboratory samples in cleanroom environments by using particle-certified glass containers.
Protect the quality of your products or lab samples. Thermo Scientific™ Particle-Certified Glass Containers are tested and certified to meet the low particle criteria; assembled to contain as few as 5 particles per milliliter > 0.5 microns. A large variant of standard particle-certified containers are available. Non-standard containers may be custom cleaned on request.
Key Features:
  • Class 100/10 glass
  • Certificate of Analysis is included with each shipment
  • Available in clear or amber
  • Assembled with low-shedding polypropylene septa with chemically inert PTFE faced liners that do not contain adhesives
  • Individual bottles double bagged to maintain integrity until point of use
  • 4 L jugs suitable for hazardous shipping in conjunction with combination packaging
Recommended for:
  • Low particle applications
TRUSTED_SUSTAINABILITY
Specifications
Capacity 16 oz., 500mL
Capacity (Metric) 500 mL
Capacity (English) 16 oz.
Color Clear
Description Clear Glass Wide Mouth Jar with Polypropylene Closure, 500 mL
Diameter (English) 3.5 in.
Material Glass
No. per Case 12
Mouth Wide
Height (Metric) 98.4 mm
Product Type Sample Jar
Height (English) 3.875 in.
Closure Liner PTFE
Closure Material Polypropylene, Low-shedding
Closure Size 89-400 mm
Diameter (Metric) 90 mm