missing translation for 'onlineSavingsMsg'
Learn More

PBD-5,11-dione, TRC

Product Code. 30413297
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Product Code. Quantity
30413297 250 mg
30409731 100 mg
30423484 500 mg
3 options
This item is not returnable. View return policy

Product Code. 30413297

Brand: Toronto Research Chemicals TRCP344510250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18877-34-4
Formula Weight 216.0899
InChI Formula InChI=1S/C12H12N2O2/c15-11-10-6-3-7-14(10)12(16)8-4-1-2-5-9(8)13-11/h1-2,4-5,10H,3,6-7H2,(H,13,15)/t10-/m0/s1
IUPAC Name (6aS)-6a,7,8,9-tetrahydro-5H-pyrrolo[2,1-c][1,4]benzodiazepine-6,11-dione
Molecular Formula C12H12N2O2
Molecular Weight (g/mol) 216.24
SMILES O=C1Nc2ccccc2C(=O)N3CCC[C@@H]13
Synonym (11aS)-2,3-Dihydro-1H-pyrrolo[2,1-c][1,4]benzodiazepine-5,11(10H,11aH)-dione, (S)-(+)-2,3-Dihydro-1H-pyrrolo[2,1-c][1,4]benzodiazepine-5,11(10H,11aH)-dione, 1,2,3,11a-Tetrahydro-5H-benzo[e]pyrrolo[1,2-a][1,4]diazepine-5,11(10H)-dione
Chemical Name or Material PBD-5,11-dione

Specifications

Quantity 250 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.