Learn More
PBS, (1X PBS Liq.), w/o Calcium and Magnesium, 100mL, MP Biomedicals
Product Code. 17398593 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. 17398593
Brand: MP Biomedicals 091860449