missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ PDE1B Recombinant Human Protein, N-His.GST Tag

Expressed from DNA sequence encoding human PDE1B long isoform (Met 1-Asp 536) fused to N-terminal polyhistidine-tagged GST tag at N-terminus

Brand:  Sino Biological™ 11746H20B5

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15269105

  • 110.00€ / 5µg


Estimated Shipment
14-08-2020
Add to basket

Description

Description

  • Formulation: Lyophilized in 50mM Tris, 100mM NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
Specifications

Specifications

Insect
Assay Standard
Lyophilized
≥90% by SDS-PAGE
Human
<1 EU/μg
PDE1B
GST Tag, His Tag
Other proteins
Analysis of protein stucture and used in cell biology research applications
89.2kd
5μg
Full Length, Recombinant
Phosphodiesterase 1B, Calmodulin-Dependent
PDE1B Recombinant Human Protein
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.