Learn More
Lenz Laborglasinstrumente™ Pear-shaped Evaporating Flask with Conical Ground Joint
Click to view available options
Volume (Metric):
10 mL
25 mL
50 mL
3000 mL
Neck Finish:
14/23
29/32
39/32
Unit Size:
Each
Pack of 10
Specifications
Specifications
| Material | Duran Glass |
| Type | Evaporating Flask |
| For Use With (Equipment) | Rotary Evaporators |
| Neck Finish | 14/23 |
| Shape | Pear |
| Volume (Metric) | 25 mL |
| Diameter (Metric) | 40 mm |