Learn More
Lenz Laborglasinstrumente™ Pear-Shaped Flasks with Conical Ground Joint
Product Code. 11572304
missing translation for 'orderingAttributeHoverText'
Diameter (Metric) Outer:
27 mm
33 mm
42 mm
51 mm
64 mm
85 mm
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
100 mL
250 mL
Socket Joint:
14/23 NS
19/26 NS
29/32 NS
missing translation for 'unitSize'
Pack of 10
Product Code. 11572304
missing translation for 'mfr': Lenz Laborglasinstrumente™ 03032928
Only null left
Add to basket
Add to basket
Especially for distillation and evaporation and provides for ease of sample removal. Lenz Laborglasinstrumente™ Pear-Shaped Flasks with Conical Ground Joint are manufactured from durable DURAN™ borosilicate glass.
  • Acc. to DIN 12383
  • With inscription label
  • Corrosion-free and resistant to thermal shock
  • With conical joint, featuring high taper accuracy and excellent tightness
Specifications
Capacity (Metric) 50 mL
Material Borosilicate Glass, Duran Glass
Socket Joint 29/32 NS
Diameter (Metric) Outer 51 mm