Learn More
Lenz Laborglasinstrumente™ Pear-Shaped Flasks with Conical Ground Joint
Product Code. p-8006062
Click to view available options
Diameter (Metric) Outer:
27 mm
33 mm
42 mm
51 mm
64 mm
85 mm
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
100 mL
250 mL
Socket Joint:
14/23 NS
19/26 NS
29/32 NS
Unit Size:
Pack of 10
Product Code. 10087780
Brand: Lenz Laborglasinstrumente™ 03031437
Only null left
Add to basket
Add to basket
Especially for distillation and evaporation and provides for ease of sample removal. Lenz Laborglasinstrumente™ Pear-Shaped Flasks with Conical Ground Joint are manufactured from durable DURAN™ borosilicate glass.
  • Acc. to DIN 12383
  • With inscription label
  • Corrosion-free and resistant to thermal shock
  • With conical joint, featuring high taper accuracy and excellent tightness
Specifications
Capacity (Metric) 100 mL
Material Borosilicate Glass, Duran Glass
Socket Joint 14/23 NS
Diameter (Metric) Outer 64 mm