Learn More
Cytiva PEEK Male Connector
Shop All Cytiva Products
Click to view available options
Connections:
1/16 in. Female to M6 Male
1/16 in. Male
1/16 in. Male to 1/16 in. Male for tubing ID 0.13 mm
1/16 in. Male to 1/16 in. Male for tubing ID 0.25 mm
3 x 5/16 in. Female
3/16 in. Male
M6 Female to 5/16 in. Male
Unit Size:
1 set
Each
Pack of 10
Pack of 2
Description
- Chromatography accessory
- Complete Packsize 2
- Select from a variety of connections sizes and internal diamters
- For use with chromatography tubing
- Connectors made of Polyetheretherketone (PEEK)
Specifications
Specifications
| Type | Male Connector |
| Connections | 1/16 in. Male |
| Material | PEEK |
| For Use With (Equipment) | 1/16 in. O.D. Tubing, 5/16 in. Male |