Learn More
Cytiva PEEK Male Connector
Connect tubing of different sizes. Cytiva Life Sciences™ PEEK Male Connector is an important accessory for optimal chromatography.
31.00€ - 464.00€
Product Code | Brand | Connections | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Connections | Price | Quantity & Availability | |||||
11515724
|
Cytiva
18-1112-57 |
1/16 in. Female to M6 Male |
149.94€
Pack of 2 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
11340062
|
Cytiva
18-1127-07 |
1/16 in. Male |
296.00€
Pack of 10 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
11729565
|
Cytiva
18-1120-92 |
1/16 in. Male to 1/16 in. Male for tubing ID 0.25 mm |
31.00€
Pack of 2 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
11370052
|
Cytiva
18-1112-49 |
3/16 in. Male |
264.00€
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
12421110
|
Cytiva
18-1120-90 |
1/16 in. Male to 1/16 in. Male for tubing ID 0.13 mm |
144.00€
1 set |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
11300072
|
Cytiva
18-1170-59 |
3 x 5/16 in. Female |
464.00€
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
11360062
|
Cytiva
18-1169-16 |
M6 Female to 5/16 in. Male |
113.00€
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
Description
- Chromatography accessory
- Complete Packsize 2
- Select from a variety of connections sizes and internal diamters
- For use with chromatography tubing
- Connectors made of Polyetheretherketone (PEEK)
Specifications
Male Connector | |
PEEK |