Learn More
Thermo Scientific™ PEEK Sleeves for Fused Silica HPLC Capillary Tubing
Product Code. 10611416 Shop All Thermo Scientific Products
Click to view available options
Description:
0.008 in. ID; Yellow
0.010 in. ID; Blμe
0.012 in. ID; Natural
0.015 in. ID; Orange
0.021 in. ID; Natural
0.030 in.ID; Natural
Unit Size:
Each
Product Code. 10611416
Brand: Thermo Scientific™ F231
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Make capillary tubing connections with Thermo Scientific™ PEEK Sleeves for Fused Silica Capillary Tubing.
Use Thermo Scientific™ PEEK Sleeves for Fused Silica Capillary Tubing to connect capillary tubing to ports designed to receive 1/16 in. O.D. tubing. Chemically inert and flexible, they are capable of withstanding high pressures.
TRUSTED_SUSTAINABILITY
Specifications
For Use With Fused Silica Capillary Tubing
Type Capillary Tubing Sleeve
Description 0.021 in. ID; Natural
Diameter (English) Inner 0.021 in.
Diameter (Metric) Inner 0.533 mm
Material PEEK
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.