missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical PEEK Tubing, 1/8 in. O.D., 100 ft
Designed specifically for modern LC, LCMS and automation useage. A flexible alternative to stainless steel tubing in high pressure applications, it ensures reproducibility in your chromatography flow rates and results through smooth internal surface along with consistent and tight toleranced IDs
625.00€
Specifications
Color | Natural |
---|---|
pH Range | 2 to 14 |
Diameter (English) Outer | 1/8 in. |
Overall Length (English) | 100 ft. |
Material | PEEK |
Product Code | Brand | Color | Diameter (English) Inner | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Diameter (English) Inner | Price | Quantity & Availability | ||||
15771039
|
Trajan Scientific and Medical
1303062000-100F |
Natural | 0.062 in. |
625.00€
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
||||
15711049
|
Trajan Scientific and Medical
1303080000-100F |
Natural | 0.080 in. |
625.00€
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
||||
Description
- Excellent chemical resistance and inert to most commonly used solvents. Exceptions are very caustic reagents, strong acids and bases
- Bio-inert ad bio-compatible
- Color coded to industry standards for easy identification of inner diameter
- Leach-free colorants
- Smooth inner walls and precise IDs
- Works well with either polymer fittings as well as stainless steel fittings
- Can be formed to specific customer needs
Specifications
Natural | |
1/8 in. | |
PEEK |
2 to 14 | |
100 ft. | |
100°C |
For research use only