missing translation for 'onlineSavingsMsg'
Learn More

Pefloxacin Mesilate Dihydrate, Mikromol™

Product Code. 17963236
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17963236 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17963236 Supplier Mikromol Supplier No. MM3495.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Pefloxacin Mesilate Dihydrate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pefloxacin methane sulfonate dihydrate
CAS 149676-40-4
Analyte or Component Names Pefloxacin methane sulfonate dihydrate
Shipping Condition Room Temperature
Molecular Formula C17 H20 F N3 O3 . C H4 O3 S . 2 H2 O
Certifications/Compliance ISO 17034
Synonym 1-Ethyl-6-fluoro-1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic acid methanesulfonate (1:1), 1-Ethyl-6-fluoro-1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic acid monomethanesulfonate, 41-982RP, Peflacin, Pefloxacin mesylate, Pefloxacin methanesulfonate
InChI Formula InChI=1S/C17H20FN3O3.CH4O3S.2H2O/c1-3-20-10-12(17(23)24)16(22)11-8-13(18)15(9-14(11)20)21-6-4-19(2)5-7-21;1-5(2,3)4;;/h8-10H,3-7H2,1-2H3,(H,23,24);1H3,(H,2,3,4);2*1H2
SMILES O.O.CCN1C=C(C(=O)O)C(=O)c2cc(F)c(cc12)N3CCN(C)CC3.CS(=O)(=O)O
IUPAC Name 1-ethyl-6-fluoro-7-(4-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid;methanesulfonic acid;dihydrate
Molecular Weight (g/mol) 465.49
Quantity 250 mg
Formula Weight 465.1581 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.