missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PEI STAR-Go™ transfection reagent
Description
PEI STAR-Go™ is a ready-to-use aqueous formulation of our high performance PEI STAR™ transfection reagent (Cat. No. 7854), provided as a sterile-filtered solution. PEI STAR™ is a chemically-defined, linear polyethylenimine (PEI) transfection reagent for affordable and scalable transient gene expression. PEI is a synthetic polymer with an exceptionally high positive charge density in pH-neutral solutions. Positively charged PEI binds strongly to negatively charged DNA and imparts a net cationic charge, allowing the DNA to enter cells. PEI is a non-viral vector commonly used to transfect HEK293, CHO, and a range of other eukaryotic cell lines. Applications include production of lentivirus and rAAV, recombinant proteins, and antibodies. Supplied as a 1mg/mL aqueous solution
PEI STAR-Go is a trademark of Bio-Techne Corp.
Specifications
Specifications
| Chemical Name or Material | Poly[imino(1,2-ethanediyl)] hydrochloride |
| CAS | 49553-93-7 |
| Quantity | 100 mL |
| Target | Transfection Reagent |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?