missing translation for 'onlineSavingsMsg'
Learn More

3M™ PELTOR™ 28 dB Earmuffs

Product Code. 15789907
Click to view available options
Model:
H31P3A 300
H31P3B 300
H31P3BB 300
H31P3E 300
H31P3E 300-GB
H31P3E 300-GU
H31P3K 300-GU
H31P3K-300
H31P3K-300-GU
Unit Size:
Pack of 20
This item is not returnable. View return policy

Product Code. 15789907

Brand: 3M™ 7000107755

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

3M™ PELTOR™ Earmuffs are ultra light and slim hearing protectors that are ideal for use in forestry, sawmills, and industry.

  • Slim hearing protector ideal for use in forestry, sawmills and industry
  • Earmuff cup has a recess at the top making it a more comfortable fit with Safety Helmets
  • Wide, comfortable sealing rings are filled with a unique combination of fluid and foam, providing optimum sealing and ideal comfort even during long periods of use
  • Earcup pivot points tilt for optimum comfort and efficiency
  • Variety of helmet attachment for helmet mounting
TRUSTED_SUSTAINABILITY

Specifications

Color Hi-Viz
For Use With (Application) Forestry, sawmills, industry
Model H31P3E 300-GB
Noise Reduction Rating (NRR) 28 dB
Product Type Helmet Mounted Ear Muffs
Dielectric No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.