Learn More
3M™ PELTOR™ Earmuffs, Green
Product Code. p-7214254
Click to view available options
Model:
X1A
X1P3E
CSA Z94.2:
Class A
Class B
Dielectric:
No
Yes
Style:
Hard Hat
Over-the-head
Unit Size:
1 pair
Each
Product Code. 13479918
Brand: 3M™ 7000103987
Only null left
Add to basket
Add to basket
3M™ PELTOR™ Earmuffs X1 are our lightweight hearing protectors with comfortable, soft ear cushions.
  • Reduces noise levels up to 27 dB
  • Wide flat headband is electrically insulted making the earmuffs electrically resistant or 'dielectric'
  • Colour-coding makes it easy to identify the level of protection being worn
  • Slim cups offer improved compatibility with other safety equipment
  • Wide soft ear cushions help workers stay comfortable
  • Twin headband design provides heat dispersion and internal stainless steel design offers consistent pressure and comfortable fit over an 8 hour period
  • Designed to optimise weight distribution and balance for outstanding comfort
  • ABS plastic cups are rigid, high strength and impact resistant
  • Thermoplastic polyurethane (TPU) soft outer cup coating protects from abrasion and bumps
Compliance
3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
TRUSTED_SUSTAINABILITY
Specifications
Color Black, Green
ANSI S3.1974 Yes
Cushion Material Foam
CSA Z94.2 Class A
Cushion Color Black
Headband Material Wire
For Use With (Application) Composite Finishing, Final Finish, Grinding, Machine Operations, Paint Preparation, Painting, Welding
Model X1A
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 22 dB
Product Type X1 Series
Headband Color Black
Dielectric Yes
Material ABS/TPU, PU Foam and PVC
Style Over-the-head