Learn More
3M™ PELTOR™ Kids Ear Muffs
Product Code. 15749057
Click to view available options
Color:
Black, Pink
Green
Unit Size:
Pack of 6
Product Code. 15749057
Brand: 3M™ 7000108439
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Specifications
Color Black, Pink
For Use With (Application) Designed For Younger Wearers, Including Children, To Protect Against Moderate Levels of Noise in Everyday Situation Such as Social and Leisure Activities.
Noise Reduction Rating (NRR) 27 dB
Plug Shape Oval
Product Type Kid Ear Muffs
Style Headband