missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ PELTOR™ Optime™ I Ear Muffs
Click to view available options
Noise Reduction Rating (NRR):
27 dB
Plug Shape:
Rounded
Unit Size:
Each
Description
- Low-profile design which minimizes obtrusiveness
- Large space inside cup helps reduce moisture and heat build-up
- Easy-to-replace cushions and inserts helps keep them hygienically clean
- Soft, wide cushions help reduce pressure around the ears and improve comfort and wearability
Specifications
Specifications
| Color | Yellow |
| Certifications/Compliance | CE Approved |
| For Use With (Application) | Used For workshop and work outdoors |
| Noise Reduction Rating (NRR) | 27 dB |
| Plug Shape | Rounded |
| Product Type | Hearing Protection Optime 1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction