missing translation for 'onlineSavingsMsg'
Learn More

3M™ PELTOR™ Optime™ III Ear Muffs

Product Code. 11739315
Click to view available options
Color:
Black, Red
Green, Black
Style:
Headband
Helmet-mounted
Neckband
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11739315

Brand: 3M™ 7000039622

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Developed for use in extremely high noise environments, particularly those dominated by high frequency sounds. 3M™ PELTOR™ Optime™ III Ear Muffs are based on technology featuring double cup design to help minimize resonance effect, resulting in excellent high frequency attenuation.

  • Modern, stylish cup design
  • Good speech intelligibility
  • Large space inside cup helps reduce moisture and heat build-up
  • Easy-to-replace cushions and inserts help keep them hygienically clean
  • Unique low-profile headband design helps maintain constant pressure
  • Soft, wide cusions help reduce pressure around the ears and improve comfort and wearability
TRUSTED_SUSTAINABILITY

Specifications

Color Black, Red
Certifications/Compliance CE Approved
For Use With (Application) Mining, Marine, Airports, and Power Station
Plug Shape Oval
Product Type Hearing Protection Optime III
Style Headband
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.