missing translation for 'onlineSavingsMsg'
Learn More

3M™ PELTOR™ X4 Series Ear Muffs

Product Code. p-7214256
Click to view available options
Model:
X4A
CSA Z94.2:
Class AL
Dielectric:
Yes
Style:
Over-the-head
Unit Size:
Each

Product Code. 13499918

Brand: 3M™ 7000103993

Only null left
Add to basket
Edge
Add to basket

PELTOR™ X4 Series Ear Muffs combines high attenuation with a sleek, low-profile and lightweight design. Available in over-the-head or hard hat-attached styles.

  • Light and small cup profile with high level of attenuation
  • High level of protection without all the bulk
  • Innovative foams and proprietary high density offers slow recovery
  • Bright yellow-green color cups help employers verify hearing protectors are being worn
  • Available in electrically insulated models (exception is model X4P3E)
  • Compatibility with hard hats (exception is model X4A)

Compliance

3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

TRUSTED_SUSTAINABILITY

Specifications

Color Black, Chartreuse
ANSI S3.1974 Yes
Cushion Material Foam
CSA Z94.2 Class AL
Cushion Color Black
Headband Material Wire
For Use With (Application) Assembly, cleaning, demolition, facility maintenance, grinding, machine operations, painting, sanding, welding
Model X4A
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 27 dB
Product Type X4 Series
Headband Color Black
Dielectric Yes
Style Over-the-head
Single Number Rating (SNR) Europe 33 dB
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.