missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl 2-thiomorpholin-4-ylisonicotinate, 97%, Thermo Scientific™

Catalog Number p-3541155
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
934570-42-0
C16H11F5N2O2S
390.33
MFCD09702363
CXWZKWSFJLNFMF-UHFFFAOYSA-N
pentafluorophenyl 2-thiomorpholin-4-ylisonicotinate, pentafluorophenyl 2-thiomorpholin-4-ylpyridine-4-carboxylate, pentafluorophenyl 2-thiomorpholin-4-yl pyridine-4-carboxylate, 2,3,4,5,6-pentakis fluoranyl phenyl 2-thiomorpholin-4-ylpyridine-4-carboxylate, 2,3,4,5,6-pentafluorophenyl 2-thiomorpholin-4-yl pyridine-4-carboxylate, 2-thiomorpholin-4-yl-4-pyridinecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester
24229510
(2,3,4,5,6-pentafluorophenyl) 2-thiomorpholin-4-ylpyridine-4-carboxylate
FC1=C(F)C(F)=C(OC(=O)C2=CC(=NC=C2)N2CCSCC2)C(F)=C1F
This item is not returnable. View return policy
934570-42-0
C16H11F5N2O2S
390.33
MFCD09702363
CXWZKWSFJLNFMF-UHFFFAOYSA-N
pentafluorophenyl 2-thiomorpholin-4-ylisonicotinate, pentafluorophenyl 2-thiomorpholin-4-ylpyridine-4-carboxylate, pentafluorophenyl 2-thiomorpholin-4-yl pyridine-4-carboxylate, 2,3,4,5,6-pentakis fluoranyl phenyl 2-thiomorpholin-4-ylpyridine-4-carboxylate, 2,3,4,5,6-pentafluorophenyl 2-thiomorpholin-4-yl pyridine-4-carboxylate, 2-thiomorpholin-4-yl-4-pyridinecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester
24229510
(2,3,4,5,6-pentafluorophenyl) 2-thiomorpholin-4-ylpyridine-4-carboxylate
FC1=C(F)C(F)=C(OC(=O)C2=CC(=NC=C2)N2CCSCC2)C(F)=C1F
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 390.33
Percent Purity 97%
Chemical Name or Material Pentafluorophenyl 2-thiomorpholin-4-ylisonicotinate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'