missing translation for 'onlineSavingsMsg'
Learn More

Pentane, AMD CHROMASOLV™, ≥99%, Honeywell Riedel-de Haën™

Catalog Number p-7111871
Click to view available options
Quantity:
2.5 L
4 x 2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
109-66-0
C5H12
72.15
MFCD00009498
OFBQJSOFQDEBGM-UHFFFAOYSA-N
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
8003
CHEBI:37830
pentane
CCCCC
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
109-66-0
C5H12
72.15
MFCD00009498
OFBQJSOFQDEBGM-UHFFFAOYSA-N
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
8003
CHEBI:37830
pentane
CCCCC

CHROMASOLV™ AMD,≥99%

  • Solvents designed for high-performance thin-layer chromatography
TRUSTED_SUSTAINABILITY
Melting Point -130°C (literature)
CAS Max % ≤100.0000%
Linear Formula CH3(CH2)3CH3
UN Number UN1265
Beilstein 969132
Formula Weight 72.15
Percent Purity >99%
Grade High Purity
Density 0.626 g/mL at 25 °C(lit.)
Boiling Point 36°C to 37°C
Quantity 2.5 L
Flash Point <−35°C
Packaging Glass Bottle
Chemical Name or Material Pentane, AMD CHROMASOLV™, ≥99%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'