missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentapiperium Metilsulfate, LoGiCal
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | Pentapiperium Metilsulfate |
| CAS | 7681-80-3 |
| Analyte or Component Names | Pentapiperium Metilsulfate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H30 N O2 . C H3 O4 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Piperidinium, 1,1-dimethyl-4-[(3-methyl-1-oxo-2-phenylpentyl)oxy]-, methyl sulfate (1:1), 4-Hydroxy-1,1-dimethylpiperidinium methyl sulfate 3-methyl-2-phenylvalerate (6CI,7CI), Piperidinium, 1,1-dimethyl-4-[(3-methyl-1-oxo-2-phenylpentyl)oxy]-, methyl sulfate (9CI), Piperidinium, 4-hydroxy-1,1-dimethyl-, methyl sulfate (salt), 3-methyl-2-phenylvalerate (8CI), 1-Methyl-4-piperidyl 3-methyl-2-phenylvalerate compound with dimethyl sulfate (1:1), 4-Hydroxy-1,1-dimethylpiperidinium methylsulfate dl-3-methyl-2-phenylvalerate, AY-5810, Crylene, Hycholin, Pentapiperide methylsulfate, Pentapiperium methylsulfate, Pentapiperium metilsulfate, Perium, Quilene |
| InChI Formula | InChI=1S/C19H30NO2.CH4O4S/c1-5-15(2)18(16-9-7-6-8-10-16)19(21)22-17-11-13-20(3,4)14-12-17;1-5-6(2,3)4/h6-10,15,17-18H,5,11-14H2,1-4H3;1H3,(H,2,3,4)/q+1;/p-1 |
| Solution Type | Neat |
| SMILES | CCC(C)C(C(=O)OC1CC[N+](C)(C)CC1)c2ccccc2.COS(=O)(=O)[O-] |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction