missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentyltrichlorosilane, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
50g
Specifications
Specifications
| CAS | 107-72-2 |
| Color | Colorless to Straw |
| Density | 1.1420g/mL |
| Boiling Point | 173°C |
| Flash Point | 31°C |
| Infrared Spectrum | Authentic |
| Packaging | AcroSeal™ Glass Bottle |
| Molecular Formula | C5H11Cl3Si |
| Refractive Index | 1.4395 to 1.4415 (20°C, 589nm) |
| Quantity | 50 g |
| Show More |
Safety and Handling
Product Identifier
- Pentyltrichlorosilane
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
- EUH014-Reacts violently with water.
- EUH029-Contact with water liberates toxic gas.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction