missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Pep 2-8
GREENER_CHOICE

Product Code. 18771429
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18771429 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18771429 Supplier Tocris Bioscience™ Supplier No. 6423/1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent PCSK9 inhibitor

Pep 2-8 is a proprotein convertase subtilisin/kexin type 9 (PCSK9) inhibitor. Potent inhibitor of PCSK9 binding to LDL receptor (IC50 = 0.8 μM). Restores LDL uptake in HepG2 cells treated with PCSK9.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1541011-97-5
Content And Storage Store at -20°C
Molecular Weight (g/mol) Observed MW: 1715.88
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.