Learn More
Gibco™ PeproGMP® Human IL-15 Recombinant Protein, PeproTech®
Description
Additional product information and specifications are provided on the Certificate of Analysis. All PeproGMP® products are manufactured using animal-derived component free materials. Product Description/Intended Use: PeproTech GMP recombinant proteins are manufactured for use as ancillary materials by applying applicable principles of GMP and quality control requirements from USP (United States Pharmacopeia) Chapter 1043 Ancillary Materials for Cell, Gene, and Tissue-Engineered Products. Product use statement: For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Specifications
| Accession Number | P40933 |
| For Use With (Application) | Control, Western Blot Control, Bioactivity |
| Format | Lyophilized |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 3600 |
| Molecular Weight (g/mol) | 12.9 kDa |
| Name | PeproGMP™ Human IL-15 |
| Purification Method | Purified |
| Quantity | 1 mg |
| Storage Requirements | -20°C or -80°C if preferred |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.