missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Pepsinogen II/PGC Human ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ EHPGC

Additional Details : Weight : 0.00400kg

 View more versions of this product

Product Code. 15455103

  • 652.00€ / 1 set
Estimated Shipment: 14-05-2024
to see stock.
Add to basket

Description

Description

The Human Pepsinogen II (Hu PGC) ELISA quantitates Hu PGC in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu PGC. Principle of the method The Human PGC solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Pepsin is one of the main proteolytic enzymes secreted by the gastric mucosa. Pepsin consists of a single polypeptide chain and arises from its precursor, pepsinogen, by removal of a 41 amino acid segment from the N-terminus. Pepsinogen is synthesized in the stomach lining, and hydrochloric acid, also produced by the gastric mucosa, is necessary to convert the inactive enzyme and to maintain the optimum acidity (pH 1-3) for pepsin function. Pepsin is particularly effective in cleaving peptide bonds involving aromatic amino acids. Pepsin shows extremely broad specificity, and although bonds involving phenylalanine and leucine are preferred, many others are also cleaved to some extent. The amino acid composition of Pepsin C differs from those of pepsinogen and pepsin especially in the content of basic amino acids, glutamic acid, aspartic acid, leucine and isoleucine.
Specifications

Specifications

P20142
40 pg/mL
Solid-phase sandwich ELISA
ELISA Kit
Plasma, Serum, Supernatant
96 assays
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
5225
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
96 Tests
Other Research
2°C to 8°C
Human
4 hr. 45 min.
40.96-10,000 pg/mL
40pg/mL to 10,000pg/mL
Biotin
Other Proteins
This ELISA pair antibody detects human PG II. Other species not determined.
Human
Colorimetric Microplate Reader
PEPC,PGII
<10%
HRP
RUO
Plasma, 10 μL; Serum, 10 μL; Supernatant, 100 μL
RP11-298J23.1, PEPC, PGII
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions