missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Peptone, Granulated
Click to view available options
Quantity:
500 g
1 kg
5 kg
Unit Size:
1kg
500g
5kg
Description
Mixtures of peptides and free amino acids from peptic digest of animal tissue.
Mixtures of peptides and free amino acids from peptic digest of animal tissue.
- Amino Nitrogen (AN): 2.5-3.5%
- Loss on Drying: <6.0%
Specifications
Specifications
| Quantity | 1 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction