missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro-1-iodohexane, 98+%, stab. with copper, Thermo Scientific Chemicals

Catalog Number p-7025474
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
355-43-1
C6F13I
445.95
MFCD00001063
BULLJMKUVKYZDJ-UHFFFAOYSA-N
perfluorohexyl iodide, perfluoro-1-iodohexane, 1-iodoperfluorohexane, perfluoro-n-hexyl iodide, tridecafluorohexyl iodide, unii-a2a17jug4r, hexane, 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-6-iodo, tridecafluoro-1-iodohexane, a2a17jug4r, hexane, tridecafluoro-6-iodo
67733
1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-6-iodohexane
C(C(C(C(F)(F)I)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F
This item is not returnable. View return policy
355-43-1
C6F13I
445.95
MFCD00001063
BULLJMKUVKYZDJ-UHFFFAOYSA-N
perfluorohexyl iodide, perfluoro-1-iodohexane, 1-iodoperfluorohexane, perfluoro-n-hexyl iodide, tridecafluorohexyl iodide, unii-a2a17jug4r, hexane, 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-6-iodo, tridecafluoro-1-iodohexane, a2a17jug4r, hexane, tridecafluoro-6-iodo
67733
1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-6-iodohexane
C(C(C(C(F)(F)I)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F

It is used as an initiator in the presence of fac-[Ir(ppy) 3 for the photoredox catalyst activated CRP of methacrylates in visible light-induced controlled radical polymerization of methacrylates with perfluoroalkyl iodide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an initiator in the presence of fac-[Ir(ppy) 3 for the photoredox catalyst activated CRP of methacrylates in visible light-induced controlled radical polymerization of methacrylates with perfluoroalkyl iodide.

Solubility
Not miscible or difficult to mix in water.

Notes
Light sensitive. Store away from light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Melting Point -45°C
Density 2.063
Boiling Point 117°C
Refractive Index 1.329
Quantity 10 g
Beilstein 1802118
Sensitivity Light sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 445.95
Percent Purity ≥98%
Chemical Name or Material Perfluoro-1-iodohexane, stabilized with copper
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'