Learn More
Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA) Acylation Reagents
Shop All Thermo Scientific Products
Click to view available options
Standard Components:
Heptafluoro-butanoic acid anhydride
Pentafluoro-propanoic acid anhydride
Trifluoroacetic acid anhydride
Quantity:
25 g
100 g
10 x 1 mL
Unit Size:
10mL
Each
Pack of 10
Description
Prepare fluoracyl derivatives for GC/MS and for flame ionization and electron-capture detection techniques with the Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA). These reagents react readily with alcohols, phenols, and amines.
- Used to prepare fluoracyl derivatives for GC/MS
- Produce stable volatile derivatives for FID and ECD techniques
Specifications
Specifications
| For Use With | GC/MS |
| Hazards | Corrosive, Flammable |
| Storage | Room Temperature |
| Quantity | 100 g |
| Standard Components | Trifluoroacetic acid anhydride |
| Standard Concentration | 98 to 100% |