Learn More
Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA) Acylation Reagents
Product Code. 11557740 Shop All Thermo Scientific Products
Click to view available options
Standard Components:
Heptafluoro-butanoic acid anhydride
Pentafluoro-propanoic acid anhydride
Trifluoroacetic acid anhydride
Quantity:
25 g
100 g
10 x 1 mL
Unit Size:
10mL
Each
Pack of 10
Product Code. 11557740
Brand: Thermo Scientific™ TS63162
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Prepare volatile and stable fluoracyl derivatives for GC/MS with the Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA).
Prepare fluoracyl derivatives for GC/MS and for flame ionization and electron-capture detection techniques with the Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA). These reagents react readily with alcohols, phenols, and amines.
  • Used to prepare fluoracyl derivatives for GC/MS
  • Produce stable volatile derivatives for FID and ECD techniques
TRUSTED_SUSTAINABILITY
Specifications
For Use With GC/MS
Storage Room Temperature
Quantity 100 g
Standard Components Heptafluoro-butanoic acid anhydride
Standard Concentration 98 to 100%